Learn More
L(-)-Camphor, 98%, Acros Organics™
Brand: Acros Organics 108190500
Code : 29
Additional Details : CAS Number : 464-48-2 Weight : 0.21000kg Transport : UN number : 2717 Chem class : 1 Pack group : III
Quantity | 50g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
L(-)-Camphor | |
Technical Grade | |
C10H16O | |
MFCD00064148 |
464-48-2 | |
98% | |
152.24 |
Specifications
L(-)-Camphor | |
64°C | |
204.0°C | |
178.0° to 180.0°C | |
464-48-2 | |
Technical Grade | |
98% | |
152.24 | |
07, 134 | |
Glass bottle |
64g/mL | |
− 32.00 (20.00°C c=10,alcohol) | |
White to yellow | |
50g | |
Authentic | |
Adhering Crystals or Crystalline Powder | |
C10H16O | |
MFCD00064148 | |
15, 1734 | |
− 32.00 |