missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Camphor, 98%, Acros Organics™

Brand:  Acros Organics 108190100

Code : 29

Additional Details : CAS Number : 464-48-2 Weight : 0.06000kg Transport : UN number : 2717 Chem class : 1 Pack group : III


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10637062

  • £40.60 / 10g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

L(-)-Camphor
Technical Grade
C10H16O
MFCD00064148
464-48-2
98%
152.24

Specifications

L(-)-Camphor
204.0°C
178.0° to 180.0°C
464-48-2
Technical Grade
98%
152.24
07, 134
Glass bottle
64°C
White to yellow
10g
Authentic
Adhering Crystals or Crystalline Powder
C10H16O
MFCD00064148
15, 1734
− 32.00
Videos
SDS