missing translation for 'onlineSavingsMsg'
Learn More

L-Bornyl acetate, 95%, ACROS Organics™

Brand:  Acros Organics 106555000

Code : 29

Additional Details : CAS Number : 5655-61-8 Weight : 0.55000kg

Quantity 500g

 View more versions of this product

Product Code. 10599800

  • £90.50 / 500g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

L-Bornyl acetate
5655-61-8
95%
196.29
95%
≥94%
C12H20O2
MFCD00867808

Specifications

total esters (GC):> 98% (as C12H20O2)
95%
Authentic
1.4620 to 1.4650 (20°C, 589nm)
Liquid
95%
C12H20O2
MFCD00867808
500g
L-Bornyl acetate
5655-61-8
Glass bottle
0.986 to 0.99 (15°/15°C)
≥94%
(20°C, 589nm) (neat) < -38°
196.29
Clear colorless
Videos
SDS