missing translation for 'onlineSavingsMsg'
Learn More

L-aspartic acid dimethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-3212789
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy
32213-95-9
C6H12ClNO4
197.615
MFCD00038878
PNLXWGDXZOYUKB-WCCKRBBISA-N
h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
2734892
dimethyl (2S)-2-aminobutanedioate;hydrochloride
COC(=O)CC(C(=O)OC)N.Cl
This item is not returnable. View return policy
32213-95-9
C6H12ClNO4
197.615
MFCD00038878
PNLXWGDXZOYUKB-WCCKRBBISA-N
h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
2734892
dimethyl (2S)-2-aminobutanedioate;hydrochloride
COC(=O)CC(C(=O)OC)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula CH3OC(O)CH2CH(NH2)C(O)OCH3·HCl
Packaging Glass Bottle
Specific Rotation +13° to +17° (20°C, 589nm) (c=1, CH3OH)
Formula Weight 197.62
Percent Purity ≥97.5%
Specific Rotation Condition +15° (20°C c=1,Chloroform)
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartic acid dimethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'