missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid di-tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 15418476
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1791-13-5
C12H24ClNO4
281.777
MFCD00034851
GVLZIMQSYQDAHB-QRPNPIFTSA-N
h-asp otbu-otbu.hcl, l-aspartic acid di-tert-butyl ester hydrochloride, s-di-tert-butyl 2-aminosuccinate hydrochloride, h-asp otbu-otbu hcl, di-tert-butyl l-aspartate hydrochloride, l-aspartic acid, bis 1,1-dimethylethyl ester, hydrochloride, l-aspartic acid di-t-butyl ester hydrochloride, h-asp obut-obut·hcl, 1,4-di-tert-butyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-otbucl
13768146
ditert-butyl (2S)-2-aminobutanedioate;hydrochloride
CC(C)(C)OC(=O)CC(C(=O)OC(C)(C)C)N.Cl
This item is not returnable. View return policy
1791-13-5
C12H24ClNO4
281.777
MFCD00034851
GVLZIMQSYQDAHB-QRPNPIFTSA-N
h-asp otbu-otbu.hcl, l-aspartic acid di-tert-butyl ester hydrochloride, s-di-tert-butyl 2-aminosuccinate hydrochloride, h-asp otbu-otbu hcl, di-tert-butyl l-aspartate hydrochloride, l-aspartic acid, bis 1,1-dimethylethyl ester, hydrochloride, l-aspartic acid di-t-butyl ester hydrochloride, h-asp obut-obut·hcl, 1,4-di-tert-butyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-otbucl
13768146
ditert-butyl (2S)-2-aminobutanedioate;hydrochloride
CC(C)(C)OC(=O)CC(C(=O)OC(C)(C)C)N.Cl

L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Beilstein 5652807
Formula Weight 281.78
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material L-Aspartic acid di-tert-butyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'