missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid 4-cyclohexyl ester, 95%, Thermo Scientific™

Catalog Number 15460539
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
112259-66-2
C10H17NO4
215.25
MFCD00190762
IJAPBBOAGRLKMH-UHFFFAOYNA-N
h-asp ochex-oh, h-asp ochx-oh, l-aspartic acid beta-cyclohexyl ester, 2s-2-amino-4-cyclohexyloxy-4-oxobutanoic acid, ambotzhaa5960, pubchem18997, l-aspartic acid b-cyclohexyl ester, h-asp ochex-oh hydrochloride salt
14283862
2-amino-4-(cyclohexyloxy)-4-oxobutanoic acid
NC(CC(=O)OC1CCCCC1)C(O)=O
This item is not returnable. View return policy
112259-66-2
C10H17NO4
215.25
MFCD00190762
IJAPBBOAGRLKMH-UHFFFAOYNA-N
h-asp ochex-oh, h-asp ochx-oh, l-aspartic acid beta-cyclohexyl ester, 2s-2-amino-4-cyclohexyloxy-4-oxobutanoic acid, ambotzhaa5960, pubchem18997, l-aspartic acid b-cyclohexyl ester, h-asp ochex-oh hydrochloride salt
14283862
2-amino-4-(cyclohexyloxy)-4-oxobutanoic acid
NC(CC(=O)OC1CCCCC1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 215.25
Percent Purity 95%
Quantity 5 g
Physical Form Solid
Chemical Name or Material L-Aspartic acid 4-cyclohexyl ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'