missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine, Anhydrous ≥98.5%, Ultrapure, Thermo Scientific Chemicals

Catalog Number 15865258
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
70-47-3
C4H8N2O3
132.119
MFCD00064401
DCXYFEDJOCDNAF-REOHCLBHSA-N
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
6267
CHEBI:17196
(2 S)-2,4-diamino-4-oxobutanoic acid
C(C(C(=O)O)N)C(=O)N
This item is not returnable. View return policy
70-47-3
C4H8N2O3
132.119
MFCD00064401
DCXYFEDJOCDNAF-REOHCLBHSA-N
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
6267
CHEBI:17196
(2 S)-2,4-diamino-4-oxobutanoic acid
C(C(C(=O)O)N)C(=O)N
Melting Point 234°C to 236°C (decomposition)
Linear Formula NH2COCH2CH(NH2)COOH
Beilstein 1723527
Packaging Plastic Bottle
Formula Weight 132.12
Percent Purity ≥98.5%
Quantity 1 kg
Physical Form Crystalline Powder
Chemical Name or Material L-Asparagine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'