missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Asp-Asp-rhodamine Trifluoroacetate Salt, Thermo Scientific™
£55.76 - £82.28
Specifications
Color | White to Off-white |
---|---|
Product Type | Detergent |
Molecular Formula | C28H25N4O9·CF3CO2H |
Formula Weight | 788.56g/mol |
Synonym | (Asp)2-R110; Rhodamine 110, bis-(L-aspartic acid amide), trifluoroacetic acid salt |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15408599
|
Thermo Scientific Alfa Aesar
J67000.!D |
0.5 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15418599
|
Thermo Scientific Alfa Aesar
J67000.M^ |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
White to Off-white | |
C28H25N4O9·CF3CO2H | |
(Asp)2-R110; Rhodamine 110, bis-(L-aspartic acid amide), trifluoroacetic acid salt | |
Lyophilized |
Detergent | |
788.56g/mol | |
-30°C to -10°C |