missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Ascorbic Acid, Powder, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™
119.70 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Specifications
Packaging | Poly Bottle |
Quantity | 100 g |
Specific Gravity | 1L = 1.65kg |
IUPAC Name | (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one |
Formula Weight | 176.12 |
Grade | BAKER ANALYZED™ Biochemical Reagent |
Physical Form | Powder |
Chemical Name or Material | L-(+)-Ascorbic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'