missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic Acid, 99+%, Extra Pure, SLR, Fisher Chemical™
48.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
48.31 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
48.31 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
1 kg
Description
L-Ascorbic Acid, 99+%, Extra Pure, SLR
Specifications
Melting Point | 190°C |
Color | White |
pH | 2.1 to 2.6 |
Packaging | Plastic powder jar |
Quantity | 100 g |
Formula Weight | 176.13g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'