missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Arginine Hydrochloride, 98+%, ACROS Organics™

Brand:  Acros Organics 105001000

Code : 29

Additional Details : CAS Number : 1119-34-2 Weight : 0.15000kg

Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10736131

  • £31.90 / 100g


EU Stock 33
Add to basket

Specifications

Chemical Identifiers

L(+)-Arginine hydrochloride
98+%
H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
MFCD00064550
98.0 to 102.0%
C6H14N4O2·HCl
210.67

Specifications

L(+)-Arginine hydrochloride
20ppm max.
0.5% max. (105°C, 2 hrs)
98.0 to 102.0%
0.5% max. (TLC)
Plastic bottle
210.67
+21.4° to +23.6° (20°C, 589nm) (c=8, 6N HCl)
0.1% max.
White
1119-34-2
Authentic
Crystals or Crystalline Powder
98+%
C6H14N4O2·HCl
H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
MFCD00064550
300ppm max.
95% min. (c=10, H2O, 430nm) 1cm cell
100g
Videos
SDS