missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Arabitol, 99%, Thermo Scientific Chemicals

Catalog Number 10493965
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7643-75-6
C5H12O5
152.15
MFCD00064290
HEBKCHPVOIAQTA-UHFFFAOYNA-N
l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
439255
CHEBI:18403
OCC(O)C(O)C(O)CO
This item is not returnable. View return policy
7643-75-6
C5H12O5
152.15
MFCD00064290
HEBKCHPVOIAQTA-UHFFFAOYNA-N
l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
439255
CHEBI:18403
OCC(O)C(O)C(O)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 99°C to 103°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 01, 531
Merck Index 10, 784
Specific Rotation Condition −12° (20°C c=2 borax soln 2%)
Specific Rotation -12
IUPAC Name (2S,4S)-pentane-1,2,3,4,5-pentol
Formula Weight 152.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L(-)-Arabitol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'