Learn More
L-AP6, Tocris Bioscience™
Selective agonist of the «quis« effect
Brand:  Tocris (Bio-Techne) 034110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487563
  • £217.80 / 10mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Selective agonist for «quis«-sensitized site.
Specifications
Chemical Identifiers
78944-89-5
211.154
L-(+)-2-Amino-6-phosphonohexanoic acid
C6H14NO5P
QIOXWRQXHFVNLV-UHFFFAOYSA-N
2927
Specifications
L-AP6
>98%
C6H14NO5P
QIOXWRQXHFVNLV-UHFFFAOYSA-N
2-amino-6-phosphonohexanoic acid
2927
78944-89-5
10mg
L-(+)-2-Amino-6-phosphonohexanoic acid
C(CCP(=O)(O)O)CC(C(=O)O)N
211.154
211.15
Videos