missing translation for 'onlineSavingsMsg'
Learn More

L-AP5, Tocris Bioscience™

Potent and selective NMDA antagonist; less active form of DL-AP5

Brand:  Tocris (Bio-Techne) 010710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13447433

  • £169.05 / 10mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NMDA antagonist. Also agonist at quisqualate-sensitized AP6 site, where it is more potent than the isomer D-AP5 .

DL Mixture and D-isomer also available.

Specifications

Chemical Identifiers

79055-67-7
197.127
l-ap5, s-2-amino-5-phosphonopentanoic acid, 2s-2-amino-5-phosphonopentanoic acid, chembl71898, 5-phosphono-l-norvaline, tocris-0107, s-2-amino-5-phosphonovaleric acid, s-2-amino-5-phosphonopentanoicacid, s-2-amino-5-phosphono-pentanoic acid, l-+-2-amino-5-phosphonopentanoic acid
(2S)-2-amino-5-phosphonopentanoic acid
C5H12NO5P
VOROEQBFPPIACJ-BYPYZUCNSA-N
1617110
C(CC(C(=O)O)N)CP(=O)(O)O

Specifications

10mg
79055-67-7
l-ap5, s-2-amino-5-phosphonopentanoic acid, 2s-2-amino-5-phosphonopentanoic acid, chembl71898, 5-phosphono-l-norvaline, tocris-0107, s-2-amino-5-phosphonovaleric acid, s-2-amino-5-phosphonopentanoicacid, s-2-amino-5-phosphono-pentanoic acid, l-+-2-amino-5-phosphonopentanoic acid
C(CC(C(=O)O)N)CP(=O)(O)O
197.127
197.13
L-AP5
C5H12NO5P
VOROEQBFPPIACJ-BYPYZUCNSA-N
(2S)-2-amino-5-phosphonopentanoic acid
1617110
>99%
Videos
Documents