missing translation for 'onlineSavingsMsg'
Learn More

L-AP4, Tocris Bioscience™

Selective group III mGlu agonist

Brand:  Tocris (Bio-Techne) 0103/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13447423

  • £189.63 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective group III metabotropic glutamate receptor agonist. Synaptic depressant. Agonist at the quisqualate-sensitized AP6 site in hippocampus.

Available as part of the Group III mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . DL Mixture and D-isomer also available.

Specifications

Chemical Identifiers

23052-81-5
183.1
l-ap4, 2s-2-amino-4-phosphonobutanoic acid, l-+-2-amino-4-phosphonobutyric acid, chembl33567, s-2-amino-4-phosphonobutyrate, l-ap-4, 3h ap4, l-2-amino-4-phosphonobutiric acid, l-1-amino-4-phosphonobutanoic acid, s-2-amino-4-phosphonobutanoic acid
(2S)-2-amino-4-phosphonobutanoic acid
C4H10NO5P
DDOQBQRIEWHWBT-VKHMYHEASA-N
179394
C(CP(=O)(O)O)C(C(=O)O)N

Specifications

10mg
23052-81-5
l-ap4, 2s-2-amino-4-phosphonobutanoic acid, l-+-2-amino-4-phosphonobutyric acid, chembl33567, s-2-amino-4-phosphonobutyrate, l-ap-4, 3h ap4, l-2-amino-4-phosphonobutiric acid, l-1-amino-4-phosphonobutanoic acid, s-2-amino-4-phosphonobutanoic acid
C(CP(=O)(O)O)C(C(=O)O)N
183.1
183.1
L-AP4
C4H10NO5P
DDOQBQRIEWHWBT-VKHMYHEASA-N
(2S)-2-amino-4-phosphonobutanoic acid
179394
>99%
Videos
Documents