missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(-)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals
23.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
Specifications
Melting Point | -10.0°C |
Color | Colorless |
Density | 0.9400g/mL |
Boiling Point | 187.0°C to 189.0°C |
Flash Point | 71°C |
Enantiomeric Excess | > 98.5% |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | C6H5CH(CH3)NH2 |
Refractive Index | 1.5250 to 1.5270 |
Show More |
Safety and Handling
Product Identifier
- L(-)-a-Methylbenzylamine
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'