missing translation for 'onlineSavingsMsg'
Learn More

L(-)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals

Catalog Number 10573891
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
2627-86-3
C8H11N
121.18
RQEUFEKYXDPUSK-ZETCQYMHSA-N
s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine
75818
CHEBI:35321
(1S)-1-phenylethanamine
CC(C1=CC=CC=C1)N
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
2627-86-3
C8H11N
121.18
RQEUFEKYXDPUSK-ZETCQYMHSA-N
s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine
75818
CHEBI:35321
(1S)-1-phenylethanamine
CC(C1=CC=CC=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY
Melting Point -10.0°C
Color Colorless
Density 0.9400g/mL
Boiling Point 187.0°C to 189.0°C
Flash Point 71°C
Enantiomeric Excess > 98.5%
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NH2
Refractive Index 1.5250 to 1.5270
Quantity 100 g
Beilstein 12,1093
Fieser 01,838; 02,271; 03,199; 05,441; 06,457; 11,411; 12,319; 13,185; 14,257; 17,276
Merck Index 14,6026
Specific Gravity 0.94
Specific Rotation (20°C, 589nm) < -39° (neat)
Solubility Information Solubility in water: 42g/L (20°C). Other solubilities: soluble in most organic solvents,miscible with alcohols and ethers
Formula Weight 121.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material L(-)-α-Methylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • L(-)-a-Methylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'