missing translation for 'onlineSavingsMsg'
Learn More

L(+)-allo-Threonine, 99%, ACROS Organics™

£30.44 - £58.37

Chemical Identifiers

CAS 28954-12-3
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064268
InChI Key AYFVYJQAPQTCCC-HRFVKAFMSA-N
Synonym l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine
PubChem CID 99289
ChEBI CHEBI:28718
IUPAC Name (2S,3S)-2-amino-3-hydroxybutanoic acid
SMILES CC(C(C(=O)O)N)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10041701
SDS View Documents
Acros Organics
198541000
100mg Glass bottle
£30.44
100mg
View alternative products
Save Up to 
»
Estimated Shipment
17-12-2020
Please to purchase this item. Need a web account? Register with us today!
10387561
SDS View Documents
Acros Organics
198542500
250mg Glass bottle
£58.37
250mg
View alternative products
Save Up to 
»
In stock, ready to ship: 4 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

28954-12-3
119.12
AYFVYJQAPQTCCC-HRFVKAFMSA-N
99289
(2S,3S)-2-amino-3-hydroxybutanoic acid
C4H9NO3
MFCD00064268
l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine
CHEBI:28718
CC(C(C(=O)O)N)O

Specifications

L(+)-allo-Threonine
28954-12-3
C4H9NO3
Authentic
04, III, 1629
Glass bottle
CC(C(C(=O)O)N)O
119.12
CHEBI:28718
+8° to +10° (20°C, 589nm) (c=2, water)
≥98.5%
White
99%
99%
CH3CH(OH)CH(NH2)CO2H
MFCD00064268
l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine
AYFVYJQAPQTCCC-HRFVKAFMSA-N
(2S,3S)-2-amino-3-hydroxybutanoic acid
99289
119.12
Crystalline Powder
+ 9.00 (20.00°C c=2,H2O)
100mg
SDS