missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine isopropyl ester hydrochloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
L-Alanine isopropyl ester hydrochloride is a reagent used in the synthesis of anti- hepatitis C virus prodrugs based on imidazotriazine and pyrrolotriazine molecules. Further, it reacts with (1R:2R)-2-Hydroxy-πnanone-3 to prepare isopropyl (1R,2R,5R)-2′-[(2-hydroxypinan-3-ylene)amino]propanoate.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
L-Alanine isopropyl ester hydrochloride is a reagent used in the synthesis of anti- hepatitis C virus prodrugs based on imidazotriazine and pyrrolotriazine molecules. Further, it reacts with (1R:2R)-2-Hydroxy-πnanone-3 to prepare isopropyl (1R,2R,5R)-2′-[(2-hydroxypinan-3-ylene)amino]propanoate.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Formula Weight | 167.63 |
Percent Purity | 95% |
Quantity | 5 g |
Chemical Name or Material | L-Alanine isopropyl ester hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'