missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine benzyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7056654
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5557-83-5
C10H14ClNO2
215.677
MFCD00054340
RLMHWGDKMJIEHH-QRPNPIFTSA-N
h-ala-obzl.hcl, l-alanine benzyl ester hydrochloride, h-ala-obzl hcl, s-benzyl 2-aminopropanoate hydrochloride, l-alanine benzyl ester hcl, benzyl l-alaninate hydrochloride, benzyl 2s-2-aminopropanoate hydrochloride, h-ala-obzlhcl, ala-obzl hcl, nh2-ala-obn hcl
12210893
benzyl (2S)-2-aminopropanoate;hydrochloride
CC(C(=O)OCC1=CC=CC=C1)N.Cl
This item is not returnable. View return policy
5557-83-5
C10H14ClNO2
215.677
MFCD00054340
RLMHWGDKMJIEHH-QRPNPIFTSA-N
h-ala-obzl.hcl, l-alanine benzyl ester hydrochloride, h-ala-obzl hcl, s-benzyl 2-aminopropanoate hydrochloride, l-alanine benzyl ester hcl, benzyl l-alaninate hydrochloride, benzyl 2s-2-aminopropanoate hydrochloride, h-ala-obzlhcl, ala-obzl hcl, nh2-ala-obn hcl
12210893
benzyl (2S)-2-aminopropanoate;hydrochloride
CC(C(=O)OCC1=CC=CC=C1)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 215.68
Percent Purity 98%
Quantity 5 g
Physical Form Powder
Chemical Name or Material L-Alanine benzyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'