missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-Alaninamide hydrochloride, 95%

Brand:  Alfa Aesar™ H63019.14

Code : NEW

Additional Details : CAS Number : 33208-99-0 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15419458

  • £80.20 / 25g


Estimated Shipment date
from Supplier 23-01-2018
Add to basket

Specifications

Chemical Identifiers

L-Alaninamide hydrochloride
124.57
Ala-NH2.HCl
33208-99-0
MFCD00066145

Specifications

L-Alaninamide hydrochloride
Solid
124.57
Ala-NH2.HCl
25g
33208-99-0
95%
MFCD00066145
212° to 217°C (dec.)
Videos
SDS