missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-798,106, Tocris Bioscience™
Potent and highly selective EP3 antagonist
Brand: Tocris (Bio-Techne) 334210
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 10mg |
---|
Description
Potent and highly selective prostanoid EP3 receptor antagonist (Ki values are 0.3, 916, > 5000 and > 5000 nM at EP3, EP4, EP1 and EP2 respectively). Attenuates sulprostone-induced inhibition of EFS-evoked twitch and contractile responses in vivo.Chemical Identifiers
244101-02-8 | |
536.44 | |
N-[(5-Bromo-2-methoxyphenyl)sulfonyl]-3-[2-(2-naphthalenylmethyl)phenyl]-2-propenamide | |
(Z)-N-(5-bromo-2-methoxyphenyl)sulfonyl-3-[2-(naphthalen-2-ylmethyl)phenyl]prop-2-enamide |
C27H22BrNO4S | |
ODTKFNUPVBULRJ-QINSGFPZSA-N | |
71311930 | |
COC1=C(C=C(C=C1)Br)S(=O)(=O)NC(=O)C=CC2=CC=CC=C2CC3=CC4=CC=CC=C4C=C3 |
Specifications
L-798, 106 | |
244101-02-8 | |
N-[(5-Bromo-2-methoxyphenyl)sulfonyl]-3-[2-(2-naphthalenylmethyl)phenyl]-2-propenamide | |
COC1=C(C=C(C=C1)Br)S(=O)(=O)NC(=O)C=CC2=CC=CC=C2CC3=CC4=CC=CC=C4C=C3 | |
536.44 | |
536.44 |
10mg | |
C27H22BrNO4S | |
ODTKFNUPVBULRJ-QINSGFPZSA-N | |
(Z)-N-(5-bromo-2-methoxyphenyl)sulfonyl-3-[2-(naphthalen-2-ylmethyl)phenyl]prop-2-enamide | |
71311930 | |
>99% |