Learn More
Tocris Bioscience™ L-755,507
Very potent and selective β3 partial agonist
Brand:  Tocris Bioscience™ 219750
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13460774
  • £618.21 / 50mg
Estimated Shipment
04-04-2019
Add to basket
Description
Description
Potent β 3 -adrenergic receptor partial agonist > 1000-fold selective over β 1 - and β 2 -adrenoceptors (EC 50 values are 0.43, 580 and > 10000 nM for activation of cloned human β 3 -, β 1 - and β 2 -adrenoceptors respectively). Stimulates lipolysis in rhesus adipocytes in vitro (EC 50 = 3.9 nM). Enhances CRISPR-mediated homology-directed repair (HDR) efficiency 2-3-fold for large fragments and ∽9-fold for point mutations, in human induced pluripotent stem cells (iPSCs).
Specifications
Specifications
L-755,507
>98
50mg
L-755,507
584.73