Learn More
Tocris Bioscience™ L-755,507
Very potent and selective β3 partial agonist
Brand:  Tocris Bioscience™ 219710
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13450774
  • £189.63 / 10mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Potent β3-adrenergic receptor partial agonist > 1000-fold selective over β1- and β2-adrenoceptors (EC50 values are 0.43, 580 and > 10000 nM for activation of cloned human β3-, β1- and β2-adrenoceptors respectively). Stimulates lipolysis in rhesus adipocytes in vitro (EC50 = 3.9 nM). Enhances CRISPR-mediated homology-directed repair (HDR) efficiency 2-3-fold for large fragments and ∽9-fold for point mutations, in human induced pluripotent stem cells (iPSCs).
Specifications
Specifications
L-755,507
>98
10mg
L-755,507
584.73