missing translation for 'onlineSavingsMsg'
Learn More

L-745,870 trihydrochloride, Tocris Bioscience™

Highly selective D4 antagonist

Brand:  Tocris (Bio-Techne) 100250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458263

  • £474.52 / 50mg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

A highly potent and selective D4 dopamine receptor antagonist; L-745,870 has Ki values of 0.51, 2300 and 960 nM for D4, D3 and D2 subtypes respectively and > 1000-fold selectivity over 5-HT2, D1 and D5 receptors.
Specifications

Chemical Identifiers

866021-03-6
436.202
l-745,870 trihydrochloride, 3-4-4-chlorophenyl piperazin-1-yl methyl-1h-pyrrolo 2,3-b pyridine trihydrochloride, 3-4-4-chlorophenyl piperazin-1-yl methyl-1h-pyrrolo 2,3-b pyridine trihydrochloride, 3-4-4-chlorophenyl piperazin-1-yl-methyl-1h-pyrrolo 2,3-b pyridine trihydrochloride, 3-4-4-chlorophenyl piperazin-1-yl-methyl-1h-pyrrolo 2,3-b pyridine tri hydrochloride
3-[[4-(4-chlorophenyl)piperazin-1-yl]methyl]-1H-pyrrolo[2,3-b]pyridine;trihydrochloride
C18H22Cl4N4
KJSOYZLYCFYXFC-UHFFFAOYSA-N
49759035
C1CN(CCN1CC2=CNC3=C2C=CC=N3)C4=CC=C(C=C4)Cl.Cl.Cl.Cl

Specifications

L-745, 870 trihydrochloride
866021-03-6
l-745,870 trihydrochloride, 3-4-4-chlorophenyl piperazin-1-yl methyl-1h-pyrrolo 2,3-b pyridine trihydrochloride, 3-4-4-chlorophenyl piperazin-1-yl methyl-1h-pyrrolo 2,3-b pyridine trihydrochloride, 3-4-4-chlorophenyl piperazin-1-yl-methyl-1h-pyrrolo 2,3-b pyridine trihydrochloride, 3-4-4-chlorophenyl piperazin-1-yl-methyl-1h-pyrrolo 2,3-b pyridine tri hydrochloride
C1CN(CCN1CC2=CNC3=C2C=CC=N3)C4=CC=C(C=C4)Cl.Cl.Cl.Cl
436.202
436.21
50mg
C18H22Cl4N4
KJSOYZLYCFYXFC-UHFFFAOYSA-N
3-[[4-(4-chlorophenyl)piperazin-1-yl]methyl]-1H-pyrrolo[2,3-b]pyridine;trihydrochloride
49759035
>99%
Videos