missing translation for 'onlineSavingsMsg'
Learn More

L-741,626, Tocris Bioscience™

High affinity D2 antagonist

Brand:  Tocris (Bio-Techne) 100350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13488263

  • £427.72 / 50mg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

A potent D2 dopamine receptor selective antagonist, with affinities of 2.4, 100 and 220 nM for D2, D3 and D4 receptors respectively. Centrally active following systemic administration in vivo.
Specifications

Chemical Identifiers

81226-60-0
340.851
3-[[4-(4-Chlorophenyl)-4-hydroxypiperidin-l-yl]methyl-1H-indole
C20H21ClN2O
LLBLNMUONVVVPG-UHFFFAOYSA-N
133633

Specifications

L-741, 626
81226-60-0
C20H21ClN2O
LLBLNMUONVVVPG-UHFFFAOYSA-N
4-(4-chlorophenyl)-1-(1H-indol-3-ylmethyl)piperidin-4-ol
133633
50mg
>98%
3-[[4-(4-Chlorophenyl)-4-hydroxypiperidin-l-yl]methyl-1H-indole
C1CN(CCC1(C2=CC=C(C=C2)Cl)O)CC3=CNC4=CC=CC=C43
340.851
340.85
Videos