missing translation for 'onlineSavingsMsg'
Learn More

L-732,138, Tocris Bioscience™

Potent and selective NK1 antagonist

Brand:  Tocris (Bio-Techne) 086850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13448083

  • £346.92 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and highly selective competitive tachykinin NK1 receptor antagonist (IC50 = 2.3 nM).
Specifications

Chemical Identifiers

148451-96-1
472.387
3,5-bis tfm bz nactrp, 3,5-bis trifluoromethyl benzyl n-acetyltryptophan, n-acetyltryptophan 3,5-bis trifluoromethyl benzyl ester, chembl22870, n-acetyl-l-trp-3,5-bistrifluoromethylbenzyl ester, 3,5-bis trifluoromethyl phenyl methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate, n-acetyl-l-tryptophan-3,5-bistrifluoromethylbenzyl ester, n-acetyl-l-tryptophan 3,5-bis trifluoromethyl benzyl ester, l-tryptophan, n-acetyl-, 3,5-bis trifluoromethyl phenyl methyl ester, tocris-0868
[3,5-bis(trifluoromethyl)phenyl]methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
C22H18F6N2O3
BYYQYXVAWXAYQC-IBGZPJMESA-N
132837
CC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)OCC3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F

Specifications

L-732, 138
C22H18F6N2O3
BYYQYXVAWXAYQC-IBGZPJMESA-N
[3,5-bis(trifluoromethyl)phenyl]methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
132837
>98%
148451-96-1
3,5-bis tfm bz nactrp, 3,5-bis trifluoromethyl benzyl n-acetyltryptophan, n-acetyltryptophan 3,5-bis trifluoromethyl benzyl ester, chembl22870, n-acetyl-l-trp-3,5-bistrifluoromethylbenzyl ester, 3,5-bis trifluoromethyl phenyl methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate, n-acetyl-l-tryptophan-3,5-bistrifluoromethylbenzyl ester, n-acetyl-l-tryptophan 3,5-bis trifluoromethyl benzyl ester, l-tryptophan, n-acetyl-, 3,5-bis trifluoromethyl phenyl methyl ester, tocris-0868
CC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)OCC3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F
472.387
472.39
50mg
Videos
Documents