missing translation for 'onlineSavingsMsg'
Learn More

L-701,324, Tocris Bioscience™

NMDA antagonist; acts at glycine site

Brand:  Tocris (Bio-Techne) 090710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13428143

  • £148.47 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

An orally active and long acting anticonvulsant with high affinity and selectivity for the glycine site on the NMDA receptor.
Specifications

Chemical Identifiers

142326-59-8
363.797
chembl31741, 7-chloro-4-hydroxy-3-3-phenoxy phenyl-2 1h-quinolone, compund 13 pmid: 8182696, 7-chloro-4-hydroxy-3-3-pheoxyphenyl-2 1h-quinolinone, 2 1h-quinolinone, 7-chloro-4-hydroxy-3-3-pheoxyphenyl, 7-chloro-2-hydroxy-3-3-phenoxyphenyl-1h-quinolin-4-one, 7-chloro-4-hydroxy-3-3-phenoxyphenyl-1h-quinolin-2-one, tocris-0907, biomol-nt_000206, d0m1wn
7-chloro-4-hydroxy-3-(3-phenoxyphenyl)-1H-quinolin-2-one
C21H14ClNO3
FLVRDMUHUXVRET-UHFFFAOYSA-N
54682505
C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=C(C4=C(C=C(C=C4)Cl)NC3=O)O

Specifications

10mg
142326-59-8
chembl31741, 7-chloro-4-hydroxy-3-3-phenoxy phenyl-2 1h-quinolone, compund 13 pmid: 8182696, 7-chloro-4-hydroxy-3-3-pheoxyphenyl-2 1h-quinolinone, 2 1h-quinolinone, 7-chloro-4-hydroxy-3-3-pheoxyphenyl, 7-chloro-2-hydroxy-3-3-phenoxyphenyl-1h-quinolin-4-one, 7-chloro-4-hydroxy-3-3-phenoxyphenyl-1h-quinolin-2-one, tocris-0907, biomol-nt_000206, d0m1wn
C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=C(C4=C(C=C(C=C4)Cl)NC3=O)O
363.797
363.8
L-701, 324
C21H14ClNO3
FLVRDMUHUXVRET-UHFFFAOYSA-N
7-chloro-4-hydroxy-3-(3-phenoxyphenyl)-1H-quinolin-2-one
54682505
>99%
Videos
Documents