missing translation for 'onlineSavingsMsg'
Learn More

L-701,324, Tocris Bioscience™

NMDA antagonist; acts at glycine site

Brand:  Tocris (Bio-Techne) 090750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13438143

  • £492.72 / 50mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

An orally active and long acting anticonvulsant with high affinity and selectivity for the glycine site on the NMDA receptor.
Specifications

Chemical Identifiers

142326-59-8
363.797
chembl31741, 7-chloro-4-hydroxy-3-3-phenoxy phenyl-2 1h-quinolone, compund 13 pmid: 8182696, 7-chloro-4-hydroxy-3-3-pheoxyphenyl-2 1h-quinolinone, 2 1h-quinolinone, 7-chloro-4-hydroxy-3-3-pheoxyphenyl, 7-chloro-2-hydroxy-3-3-phenoxyphenyl-1h-quinolin-4-one, 7-chloro-4-hydroxy-3-3-phenoxyphenyl-1h-quinolin-2-one, tocris-0907, biomol-nt_000206, d0m1wn
7-chloro-4-hydroxy-3-(3-phenoxyphenyl)-1H-quinolin-2-one
C21H14ClNO3
FLVRDMUHUXVRET-UHFFFAOYSA-N
54682505
C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=C(C4=C(C=C(C=C4)Cl)NC3=O)O

Specifications

L-701, 324
142326-59-8
chembl31741, 7-chloro-4-hydroxy-3-3-phenoxy phenyl-2 1h-quinolone, compund 13 pmid: 8182696, 7-chloro-4-hydroxy-3-3-pheoxyphenyl-2 1h-quinolinone, 2 1h-quinolinone, 7-chloro-4-hydroxy-3-3-pheoxyphenyl, 7-chloro-2-hydroxy-3-3-phenoxyphenyl-1h-quinolin-4-one, 7-chloro-4-hydroxy-3-3-phenoxyphenyl-1h-quinolin-2-one, tocris-0907, biomol-nt_000206, d0m1wn
C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=C(C4=C(C=C(C=C4)Cl)NC3=O)O
363.797
363.8
50mg
C21H14ClNO3
FLVRDMUHUXVRET-UHFFFAOYSA-N
7-chloro-4-hydroxy-3-(3-phenoxyphenyl)-1H-quinolin-2-one
54682505
>99%
Videos