missing translation for 'onlineSavingsMsg'
Learn More

L-701,252, Tocris Bioscience™

NMDA antagonist, acts glycine site

Brand:  Tocris (Bio-Techne) 070510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13447883

  • £116.13 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

An antagonist at the glycine-NMDA site (IC50 = 420 nM). Also a potent systemic anticonvulsant.
Specifications

Chemical Identifiers

151057-13-5
263.677
7-chloro-3-cyclopropanecarbonyl-4-hydroxyquinolin-2 1h-one, 7-chloro-3-cyclopropyl-carbonyl-4-hydroxy-2 1h-quinolinone, 7-chloro-3-cyclopropylcarbonyl-4-hydroxy-2 1h-quinolinone, tocris-0705, biomol-nt_000184, 2 1h-quinolinone,7-chloro-3-cyclopropylcarbonyl-4-hydroxy, 7-chloranyl-3-cyclopropylcarbonyl-2-oxidanyl-1h-quinolin-4-one, 7-chloro-3-cyclopropanecarbonyl-2-hydroxy-1h-quinolin-4-one, 7-chloro-3-cyclopropyl-carbonyl-4-hydroxy-2 1h quinolinone, 7-chloro-3-cyclopropylcarbonyl-4-hydroxy-2 1h-quinoline
7-chloro-3-(cyclopropanecarbonyl)-4-hydroxy-1H-quinolin-2-one
C13H10ClNO3
MXEFWCFPCLDOOG-UHFFFAOYSA-N
54687453
C1CC1C(=O)C2=C(C3=C(C=C(C=C3)Cl)NC2=O)O

Specifications

10mg
151057-13-5
7-chloro-3-cyclopropanecarbonyl-4-hydroxyquinolin-2 1h-one, 7-chloro-3-cyclopropyl-carbonyl-4-hydroxy-2 1h-quinolinone, 7-chloro-3-cyclopropylcarbonyl-4-hydroxy-2 1h-quinolinone, tocris-0705, biomol-nt_000184, 2 1h-quinolinone,7-chloro-3-cyclopropylcarbonyl-4-hydroxy, 7-chloranyl-3-cyclopropylcarbonyl-2-oxidanyl-1h-quinolin-4-one, 7-chloro-3-cyclopropanecarbonyl-2-hydroxy-1h-quinolin-4-one, 7-chloro-3-cyclopropyl-carbonyl-4-hydroxy-2 1h quinolinone, 7-chloro-3-cyclopropylcarbonyl-4-hydroxy-2 1h-quinoline
C1CC1C(=O)C2=C(C3=C(C=C(C=C3)Cl)NC2=O)O
263.677
263.68
L-701, 252
C13H10ClNO3
MXEFWCFPCLDOOG-UHFFFAOYSA-N
7-chloro-3-(cyclopropanecarbonyl)-4-hydroxy-1H-quinolin-2-one
54687453
>99%
Videos
Documents