missing translation for 'onlineSavingsMsg'
Learn More

L-694,247, Tocris Bioscience™

5-HT1D agonist

Brand:  Tocris (Bio-Techne) 078150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13487993

  • £664.29 / 50mg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Selective and extremely potent 5-HT1D receptor agonist.
Specifications

Chemical Identifiers

137403-12-4
411.48
2-[5-[3-(4-Methylsulfonylamino)benzyl-1, 2, 4-oxadiazol-5-yl]-1H-indol-3-yl]ethanamine
C20H21N5O3S
HKXMQLISPYELRD-UHFFFAOYSA-N
132059

Specifications

L-694, 247
137403-12-4
C20H21N5O3S
HKXMQLISPYELRD-UHFFFAOYSA-N
N-[4-[[5-[3-(2-aminoethyl)-1H-indol-5-yl]-1,2,4-oxadiazol-3-yl]methyl]phenyl]methanesulfonamide
132059
50mg
>98%
2-[5-[3-(4-Methylsulfonylamino)benzyl-1, 2, 4-oxadiazol-5-yl]-1H-indol-3-yl]ethanamine
CS(=O)(=O)NC1=CC=C(C=C1)CC2=NOC(=N2)C3=CC4=C(C=C3)NC=C4CCN
411.48
411.48
Videos