Learn More
L-690,330, Tocris Bioscience™
Inositol monophosphatase inhibitor
Brand:  Tocris (Bio-Techne) 068110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13467943
  • £176.40 / 10mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
A potent inhibitor of inositol monophophatase; stable to hydrolysis. Induces autophagy in COS-7 cells independently of mTOR inhibition.
Specifications
Chemical Identifiers
142523-38-4
298.124
chembl34819, 1-4-hydroxyphenoxy-1-phosphonoethyl phosphonic acid, 1-4-hydroxyphenoxy-1-phosphonoethylphosphonic acid, 1-p-hydroxyphenoxy ethylenebisphosphonate, d06jat, 1-4-hydroxyphenoxy ethylenebisphosphonic acid, 1-4-hydroxyphenoxy ethylidene bisphosphonic acid, phosphonic acid, 1-4-hydroxyphenoxy ethylidene bis, 1-4-hydroxy-phenoxy-1-phosphono-ethyl-phosphonic acid, 1-4-hydroxyphenoxy-1,1-ethanediyl bis phosphonic acid
[1-(4-hydroxyphenoxy)-1-phosphonoethyl]phosphonic acid
C8H12O8P2
JKOCAAWWDVHWKB-UHFFFAOYSA-N
132449
CC(OC1=CC=C(C=C1)O)(P(=O)(O)O)P(=O)(O)O
Specifications
L-690, 330
142523-38-4
chembl34819, 1-4-hydroxyphenoxy-1-phosphonoethyl phosphonic acid, 1-4-hydroxyphenoxy-1-phosphonoethylphosphonic acid, 1-p-hydroxyphenoxy ethylenebisphosphonate, d06jat, 1-4-hydroxyphenoxy ethylenebisphosphonic acid, 1-4-hydroxyphenoxy ethylidene bisphosphonic acid, phosphonic acid, 1-4-hydroxyphenoxy ethylidene bis, 1-4-hydroxy-phenoxy-1-phosphono-ethyl-phosphonic acid, 1-4-hydroxyphenoxy-1,1-ethanediyl bis phosphonic acid
CC(OC1=CC=C(C=C1)O)(P(=O)(O)O)P(=O)(O)O
298.124
298.13
10mg
C8H12O8P2
JKOCAAWWDVHWKB-UHFFFAOYSA-N
[1-(4-hydroxyphenoxy)-1-phosphonoethyl]phosphonic acid
132449
>98%