Learn More
Tocris Bioscience™ L-689,560
Brand:  Tocris Bioscience™ 0742/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13487933
  • £170.28 / 10mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Very potent antagonist at the glycine-NMDA site. Also available as part of the NMDA Receptor - Glycine Site Tocriset™ .
Specifications
Specifications
trans-2-Carboxy-5,7-dichloro-4-phenylaminocarbonylamino-1,2,3,4-tetrahydroquinoline
>99%
Observed MW: 380.23
Soluble to 25mM in DMSO
139051-78-8
C17H15Cl2N3O3