missing translation for 'onlineSavingsMsg'
Learn More

L-655,240, Tocris Bioscience™

Potent, selective thromboxane A2/prostaglandin endoperoxide antagonist. Restrictions apply. See special conditions on product page.

Brand:  Tocris (Bio-Techne) 169850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13488653

  • £539.52 / 50mg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Potent and selective thromboxane A2/prostaglandin endoperoxide receptor antagonist. pA2 values are 8 - 8.4 in guinea pig smooth muscle; IC50 = 7 nM for inhibition of human platelet aggregation. Orally active in vivo.
Specifications

Chemical Identifiers

103253-15-2
373.852
1-[(4-Chlorophenyl)methyl]-5-fluoro-α, α, 3-trimethyl-1H-indole-2-propanoic acid
C21H21ClFNO2
JLPYLHLUHJOPNL-UHFFFAOYSA-N
122070

Specifications

L-655, 240
103253-15-2
C21H21ClFNO2
JLPYLHLUHJOPNL-UHFFFAOYSA-N
3-[1-[(4-chlorophenyl)methyl]-5-fluoro-3-methylindol-2-yl]-2,2-dimethylpropanoic acid
122070
50mg
>99%
1-[(4-Chlorophenyl)methyl]-5-fluoro-α, α, 3-trimethyl-1H-indole-2-propanoic acid
CC1=C(N(C2=C1C=C(C=C2)F)CC3=CC=C(C=C3)Cl)CC(C)(C)C(=O)O
373.852
373.85
Videos