missing translation for 'onlineSavingsMsg'
Learn More

L-651,582, Tocris Bioscience™

Ca2+ channel blocker

Brand:  Tocris (Bio-Techne) 308650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13462364

  • £705.24 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Orally active Ca2+ channel blocker; inhibits K+- and carbachol-stimulated Ca2+ influx (IC50 values are 500 and 935 nM respectively). Exhibits antiproliferative, antiangiogenic and antimetastatic activity in vivo and displays selectivity towards numerous mismatch repair-deficient tumor cell lines in vitro.
Specifications

Chemical Identifiers

99519-84-3
424.666
carboxyamidotriazole, carboxyamido-triazole, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl benzyl-1h-1,2,3-triazole-4-carboxamide, cai, 4-cai, unii-6st3zf52wb, 6st3zf52wb, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl-1h-1,2,3-triazole-4-carboxamide
5-amino-1-[[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]methyl]triazole-4-carboxamide
C17H12Cl3N5O2
WNRZHQBJSXRYJK-UHFFFAOYSA-N
108144
C1=CC(=CC=C1C(=O)C2=C(C=C(C=C2Cl)CN3C(=C(N=N3)C(=O)N)N)Cl)Cl

Specifications

L-651, 582
99519-84-3
carboxyamidotriazole, carboxyamido-triazole, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl benzyl-1h-1,2,3-triazole-4-carboxamide, cai, 4-cai, unii-6st3zf52wb, 6st3zf52wb, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl-1h-1,2,3-triazole-4-carboxamide
C1=CC(=CC=C1C(=O)C2=C(C=C(C=C2Cl)CN3C(=C(N=N3)C(=O)N)N)Cl)Cl
424.666
424.67
50mg
C17H12Cl3N5O2
WNRZHQBJSXRYJK-UHFFFAOYSA-N
5-amino-1-[[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]methyl]triazole-4-carboxamide
108144
>98%
Videos