missing translation for 'onlineSavingsMsg'
Learn More

L-4-chlorophenylalanine, 98%, Thermo Scientific™

Catalog Number p-3757261
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
14173-39-8
C9H10ClNO2
199.63
MFCD00063065,MFCD00079675
NIGWMJHCCYYCSF-UHFFFAOYNA-N
4-chloro-l-phenylalanine, l-4-chlorophenylalanine, h-phe 4-cl-oh, s-2-amino-3-4-chlorophenyl propanoic acid, l-p-chlorophenylalanine, 2s-2-amino-3-4-chlorophenyl propanoic acid, fenclonine, s, l-pcpa, unii-snm151oe2c, l-phe 4-cl-oh
736190
NC(CC1=CC=C(Cl)C=C1)C(O)=O
This item is not returnable. View return policy
14173-39-8
C9H10ClNO2
199.63
MFCD00063065,MFCD00079675
NIGWMJHCCYYCSF-UHFFFAOYNA-N
4-chloro-l-phenylalanine, l-4-chlorophenylalanine, h-phe 4-cl-oh, s-2-amino-3-4-chlorophenyl propanoic acid, l-p-chlorophenylalanine, 2s-2-amino-3-4-chlorophenyl propanoic acid, fenclonine, s, l-pcpa, unii-snm151oe2c, l-phe 4-cl-oh
736190
NC(CC1=CC=C(Cl)C=C1)C(O)=O
Melting Point 235.0°C to 238.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4CH2CH(NH2)CO2H
Quantity 100 mg
Specific Rotation − 27.00
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in ethanol
IUPAC Name (2S)-2-amino-3-(4-chlorophenyl)propanoic acid
Formula Weight 199.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-4-Chlorophenylalanine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'