missing translation for 'onlineSavingsMsg'
Learn More

L-364,373, Tocris Bioscience™

KV7.1 channel activator, activates IKs

Brand:  Tocris (Bio-Techne) 266010

Code : Z1

Additional Details : Weight : 0.00001kg

Quantity 10mg

 View more versions of this product

Product Code. 13441674

  • £188.76 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Activator of KV7.1 (KCNQ1) channels. Activates the cardiac IKs current that decreases action potential duration (APD) in cardiac myocytes.
Specifications

Chemical Identifiers

103342-82-1
397.453
r-l3, cck antagonist synthetic 11, 3r-5-2-fluorophenyl-3-1h-indol-3-ylmethyl-1-methyl-3h-1,4-benzodiazepin-2-one, d05gqk, 5-2-fluorophenyl-1,3-dihydro-3-1h-indol-3-ylmethyl-1-methyl-2h-1,4-benzodiazepin-2-one
CHEBI:34942
CN1C2=CC=CC=C2C(=NC(C1=O)CC3=CNC4=CC=CC=C43)C5=CC=CC=C5F
C25H20FN3O
CGBANSGENFERAT-JOCHJYFZSA-N
656755
(3R)-5-(2-fluorophenyl)-3-(1H-indol-3-ylmethyl)-1-methyl-3H-1,4-benzodiazepin-2-one

Specifications

L-364, 373
103342-82-1
r-l3, cck antagonist synthetic 11, 3r-5-2-fluorophenyl-3-1h-indol-3-ylmethyl-1-methyl-3h-1,4-benzodiazepin-2-one, d05gqk, 5-2-fluorophenyl-1,3-dihydro-3-1h-indol-3-ylmethyl-1-methyl-2h-1,4-benzodiazepin-2-one
CN1C2=CC=CC=C2C(=NC(C1=O)CC3=CNC4=CC=CC=C43)C5=CC=CC=C5F
397.453
CHEBI:34942
>98%
10mg
C25H20FN3O
CGBANSGENFERAT-JOCHJYFZSA-N
(3R)-5-(2-fluorophenyl)-3-(1H-indol-3-ylmethyl)-1-methyl-3H-1,4-benzodiazepin-2-one
656755
397.44
Videos