missing translation for 'onlineSavingsMsg'
Learn More

L-364,373, Tocris Bioscience™

KV7.1 channel activator, activates IKs

Brand:  Tocris (Bio-Techne) 266050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13451674

  • £848.19 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Activator of KV7.1 (KCNQ1) channels. Activates the cardiac IKs current that decreases action potential duration (APD) in cardiac myocytes.
Specifications

Chemical Identifiers

103342-82-1
397.453
r-l3, cck antagonist synthetic 11, 3r-5-2-fluorophenyl-3-1h-indol-3-ylmethyl-1-methyl-3h-1,4-benzodiazepin-2-one, d05gqk, 5-2-fluorophenyl-1,3-dihydro-3-1h-indol-3-ylmethyl-1-methyl-2h-1,4-benzodiazepin-2-one
CHEBI:34942
CN1C2=CC=CC=C2C(=NC(C1=O)CC3=CNC4=CC=CC=C43)C5=CC=CC=C5F
C25H20FN3O
CGBANSGENFERAT-JOCHJYFZSA-N
656755
(3R)-5-(2-fluorophenyl)-3-(1H-indol-3-ylmethyl)-1-methyl-3H-1,4-benzodiazepin-2-one

Specifications

50mg
103342-82-1
r-l3, cck antagonist synthetic 11, 3r-5-2-fluorophenyl-3-1h-indol-3-ylmethyl-1-methyl-3h-1,4-benzodiazepin-2-one, d05gqk, 5-2-fluorophenyl-1,3-dihydro-3-1h-indol-3-ylmethyl-1-methyl-2h-1,4-benzodiazepin-2-one
CN1C2=CC=CC=C2C(=NC(C1=O)CC3=CNC4=CC=CC=C43)C5=CC=CC=C5F
397.453
CHEBI:34942
>98%
L-364, 373
C25H20FN3O
CGBANSGENFERAT-JOCHJYFZSA-N
(3R)-5-(2-fluorophenyl)-3-(1H-indol-3-ylmethyl)-1-methyl-3H-1,4-benzodiazepin-2-one
656755
397.44
Videos
Documents