missing translation for 'onlineSavingsMsg'
Learn More

L(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3779431
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20312-36-1
C9H9O3
165.17
MFCD00004244
VOXXWSYKYCBWHO-QMMMGPOBSA-M
l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid
444718
CHEBI:43065
(2S)-2-hydroxy-3-phenylpropanoic acid
O[C@@H](CC1=CC=CC=C1)C([O-])=O
This item is not returnable. View return policy
20312-36-1
C9H9O3
165.17
MFCD00004244
VOXXWSYKYCBWHO-QMMMGPOBSA-M
l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid
444718
CHEBI:43065
(2S)-2-hydroxy-3-phenylpropanoic acid
O[C@@H](CC1=CC=CC=C1)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 121°C to 125°C
Color Beige-Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH2CH(OH)CO2H
Quantity 1 g
Specific Rotation Condition −18.50° (24°C c=1,CH3COOH)
Specific Rotation -18.5
Solubility Information Solubility in water: soluble
Formula Weight 166.18
Percent Purity 98%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material L(-)-3-Phenyllactic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'