missing translation for 'onlineSavingsMsg'
Learn More

L(-)-2-Octanol, 99+%, Thermo Scientific Chemicals

Catalog Number 10571991
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5978-70-1
C8H18O
130.23
MFCD00064284
SJWFXCIHNDVPSH-MRVPVSSYSA-N
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
80080
CHEBI:37871
(2R)-octan-2-ol
CCCCCCC(C)O
This item is not returnable. View return policy
5978-70-1
C8H18O
130.23
MFCD00064284
SJWFXCIHNDVPSH-MRVPVSSYSA-N
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
80080
CHEBI:37871
(2R)-octan-2-ol
CCCCCCC(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8300g/mL
Boiling Point 175.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5CH(OH)CH3
Refractive Index 1.4250 to 1.4270
Quantity 5 g
Beilstein 01, IV, 1770
Merck Index 15, 6841
Specific Gravity 0.83
Specific Rotation Condition −9.50° (20°C neat)
Specific Rotation − 9.50
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol and ether
Formula Weight 130.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material L(-)-2-Octanol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • L(-)-2-Octanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'