missing translation for 'onlineSavingsMsg'
Learn More

L-165,041, Tocris Bioscience™

Potent PPARδ agonist

Brand:  Tocris (Bio-Techne) 185650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13440134

  • £600.29 / 50mg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent PPARδ agonist (Ki = 6 nM); displays > 100-fold selectivity for both mouse and human PPARδ receptors over other subtypes. Raises plasma cholesterol levels in vivo, in insulin-resistant db/db mice. Neuroprotective in models of cerebral infarction and Parkinson's disease.
Specifications

Chemical Identifiers

79558-09-1
402.443
2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy acetic acid, 4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy-acetic acid, 4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxyacetic acid, 2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy-propoxy phenoxy acetic acid, 2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy acetic acid, 4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy acetic acid, 4-3-2-propyl-3-hydroxy-4-acetyl phenoxy propyloxyphenoxy-acetic acid, aceticacid, 2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy, pubchem19280, lopac-l-2167
2-[4-[3-(4-acetyl-3-hydroxy-2-propylphenoxy)propoxy]phenoxy]acetic acid
C22H26O7
HBBVCKCCQCQCTJ-UHFFFAOYSA-N
6603901
CCCC1=C(C=CC(=C1O)C(=O)C)OCCCOC2=CC=C(C=C2)OCC(=O)O

Specifications

L-165, 041
79558-09-1
2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy acetic acid, 4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy-acetic acid, 4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxyacetic acid, 2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy-propoxy phenoxy acetic acid, 2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy acetic acid, 4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy acetic acid, 4-3-2-propyl-3-hydroxy-4-acetyl phenoxy propyloxyphenoxy-acetic acid, aceticacid, 2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy, pubchem19280, lopac-l-2167
CCCC1=C(C=CC(=C1O)C(=O)C)OCCCOC2=CC=C(C=C2)OCC(=O)O
402.443
402.44
50mg
C22H26O7
HBBVCKCCQCQCTJ-UHFFFAOYSA-N
2-[4-[3-(4-acetyl-3-hydroxy-2-propylphenoxy)propoxy]phenoxy]acetic acid
6603901
>98%
Videos