missing translation for 'onlineSavingsMsg'
Learn More

L-152,804, Tocris Bioscience™

Potent, selective non-peptide NPY Y5 receptor antagonist

Brand:  Tocris (Bio-Techne) 138210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13439273

  • £163.17 / 10mg

Estimated Shipment date
from Supplier 23-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective non-peptide neuropeptide Y Y5 receptor antagonist (Ki = 26 nM for hY5). Displays > 300-fold selectivity over hY1, hY2, and hY4 receptors. Causes weight loss in diet-induced obese mice by modulating food intake and energy expenditure. Centrally active upon oral administration in vivo.
Specifications

Chemical Identifiers

6508-43-6
366.457
2-3,3-dimethyl-1-oxo-2,3,4,9-tetrahydro-1h-xanthen-9-yl-5,5-dimethylcyclohexane-1,3-dione, 2-3,3-dimethyl-1-oxo-4,9-dihydro-2h-xanthen-9-yl-5,5-dimethylcyclohexane-1,3-dione, 5,5-dimethyl-2-2,3,4,9-tetrahydro-3,3-dimethyl-1oxo-1h-xanthen-9-yl-1,3-cyclohexanedione, enamine_001564, 1,3-cyclohexanedione, 5,5-dimethyl-2-1,2,3,4-tetrahydro-3,3-dimethyl-1-oxoxanthen-9-yl, 9-4,4-dimethyl-2,6-dioxocyclohexyl-1,2,3,4-tetrahydro-3,3-dimethyl-1-xanthenone, 1,3-cyclohexanedione, 5,5-dimethyl-2-2,3,4,9-tetrahydro-3,3-dimethyl-1-oxo-1h-xanthen-9-yl, 2-3,3-dimethyl-1-oxo-2,3,4,9-tetrahydro-1h-xanthen-9-yl-5,5-dimethyl-cyclohexa, 9-2,6-dioxo-4,4-dimethylcyclohexyl-3,3-dimethyl-3,4-dihydro-9h-xanthene-1 2h-one, 2-3,3-dimethyl-1-oxo-2,3,4,9-tetrahydro-1h-xanthen-9-yl-5,5-dimethyl-cyclohexane-1,3-dione
2-(3,3-dimethyl-1-oxo-4,9-dihydro-2H-xanthen-9-yl)-5,5-dimethylcyclohexane-1,3-dione
C23H26O4
XQDXIHYKAGHZRX-UHFFFAOYSA-N
3765615
CC1(CC(=O)C(C(=O)C1)C2C3=CC=CC=C3OC4=C2C(=O)CC(C4)(C)C)C

Specifications

10mg
6508-43-6
2-3,3-dimethyl-1-oxo-2,3,4,9-tetrahydro-1h-xanthen-9-yl-5,5-dimethylcyclohexane-1,3-dione, 2-3,3-dimethyl-1-oxo-4,9-dihydro-2h-xanthen-9-yl-5,5-dimethylcyclohexane-1,3-dione, 5,5-dimethyl-2-2,3,4,9-tetrahydro-3,3-dimethyl-1oxo-1h-xanthen-9-yl-1,3-cyclohexanedione, enamine_001564, 1,3-cyclohexanedione, 5,5-dimethyl-2-1,2,3,4-tetrahydro-3,3-dimethyl-1-oxoxanthen-9-yl, 9-4,4-dimethyl-2,6-dioxocyclohexyl-1,2,3,4-tetrahydro-3,3-dimethyl-1-xanthenone, 1,3-cyclohexanedione, 5,5-dimethyl-2-2,3,4,9-tetrahydro-3,3-dimethyl-1-oxo-1h-xanthen-9-yl, 2-3,3-dimethyl-1-oxo-2,3,4,9-tetrahydro-1h-xanthen-9-yl-5,5-dimethyl-cyclohexa, 9-2,6-dioxo-4,4-dimethylcyclohexyl-3,3-dimethyl-3,4-dihydro-9h-xanthene-1 2h-one, 2-3,3-dimethyl-1-oxo-2,3,4,9-tetrahydro-1h-xanthen-9-yl-5,5-dimethyl-cyclohexane-1,3-dione
CC1(CC(=O)C(C(=O)C1)C2C3=CC=CC=C3OC4=C2C(=O)CC(C4)(C)C)C
366.457
366.46
L-152, 804
C23H26O4
XQDXIHYKAGHZRX-UHFFFAOYSA-N
2-(3,3-dimethyl-1-oxo-4,9-dihydro-2H-xanthen-9-yl)-5,5-dimethylcyclohexane-1,3-dione
3765615
>99%
Videos
Documents