missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ KYNU Recombinant Mouse Protein, His Tag

Expressed from DNA sequence encoding mouse KYNU (Q9CXF0) (Met 1-Ser 464) fused to C-terminal polyhistidine tag

Brand:  Sino Biological™ 50310M08B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15313096

  • £595.46 / 50µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH 8.0
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
70789
≥90% by SDS-PAGE
Mouse
<1 EU/μg
KYNU
Other Proteins
KYNU Recombinant Mouse Protein
Lyophilized
53.7kd
50μg
Full Length, Recombinant
Kynureninase (L-Kynurenine Hydrolase)
Assay Standard
Insect
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.