missing translation for 'onlineSavingsMsg'
Learn More

KW 3902, Tocris Bioscience™

Selective A1 antagonist

Brand:  Tocris (Bio-Techne) 416750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444274

  • £487.52 / 50mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective adenosine A1 receptor antagonist; displays 890-fold selectivity for rat A1 receptors over A2A receptors (Ki values are 0.19 and 170 nM respectively). Displays no effect on recombinant rat A3 receptors expressed on CHO cells at concentrations up to 10 μM. Exhibits diuretic and renal protective effects in rats.
Specifications

Chemical Identifiers

136199-02-5
356.47
rolofylline, 1,3-dnax, 1,3-dipropyl-8-3-noradamantyl xanthine, rolofylline usan, 8-3-noradamantyl-1,3-dipropylxanthine, chembl52333, 8-hexahydro-2,5-methanopentalen-3a 1h-yl-3,7-dihydro-1,3-dipropyl-1h-purine-2,6-dione, 8-noradamantan-3-yl-1,3-dipropylxanthine, 8-hexahydro-2,5-methanopentalen-3a 1h-yl-1,3-dipropyl-3,7-dihydro-1h-purine-2,6-dione, d0x0iy
CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C34CC5CC(C3)CC4C5
C20H28N4O2
PJBFVWGQFLYWCB-UHFFFAOYSA-N
64627

Specifications

KW 3902
136199-02-5
rolofylline, 1,3-dnax, 1,3-dipropyl-8-3-noradamantyl xanthine, rolofylline usan, 8-3-noradamantyl-1,3-dipropylxanthine, chembl52333, 8-hexahydro-2,5-methanopentalen-3a 1h-yl-3,7-dihydro-1,3-dipropyl-1h-purine-2,6-dione, 8-noradamantan-3-yl-1,3-dipropylxanthine, 8-hexahydro-2,5-methanopentalen-3a 1h-yl-1,3-dipropyl-3,7-dihydro-1h-purine-2,6-dione, d0x0iy
CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C34CC5CC(C3)CC4C5
64627
>99%
50mg
C20H28N4O2
PJBFVWGQFLYWCB-UHFFFAOYSA-N
356.47
356.46
Videos