missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kremen-2 Mouse anti-Human, Clone: 276709, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Kremen-2 |
---|---|
Clone | 276709 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Kremen-2 Gln19-Ala364 Accession # Q8NCW0 |
Gene ID (Entrez) | 79412 |
Gene Accession No. | Q8NCW0 |
Description
Kremen-2 Monoclonal specifically detects Kremen-2 in Human samples. It is validated for Western Blot.Specifications
Kremen-2 | |
Mouse myeloma cell line NS0-derived recombinant human Kremen-2 Gln19-Ala364 Accession # Q8NCW0 | |
Q8NCW0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
276709 | |
79412 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Kremen-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Kremen-1 or recombinant mouse Kremen-2 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only