Learn More
DWK Life Sciences Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
Brand:  DWK Life Sciences 774261-0014
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 12859518
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
  • Plug-type stopper with sleeve extension fits into the neck of the apparatus for a secure seal
  • Diaphragm can be punctured with a syringe needle
  • Use for outgassing NMR sample tubes, as a pressure vent or for adding reactant via a syringe
Specifications
Specifications
White
1.4 cm
NMR tubes, pressure venting or adding reactants via syringe
Plug Type Sleeve Stopper
Rubber
14/20