missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Novagen™ KOD XL DNA Polymerase

For PCR amplification of long strand and GC-rich DNA templates

Brand:  EMD Millipore™ 71087-3

Code : Z3

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12858435

  • £396.00 / Each


Estimated Shipment
01-06-2018
Add to basket

Description

Description

  • Used for amplification of large DNA fragments from purified DNA or crude samples
  • Amplifies DNA templates up to 30 kbp
  • Efficiently incorporates derivatized dNTPs
Specifications

Specifications

2.5U/μL
Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli
KOD XL DNA Polymerase
KOD Hot Start DNA Polymerase
For Accurate PCR Amplification
–20°C
250U
Videos

For Research Use Only