Learn More
Merck Millipore Novagen™ KOD Hot Start DNA Polymerase
For PCR amplification of long strand and GC-rich DNA templates, cloning and cDNA amplification applications
Brand: Merck Millipore 71086-3
Code : Z3
Additional Details : Weight : 0.01000kg
Description
- High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
- Amplifies genomic DNA templates up to 12 kbp
- Amplifies plasmid and lambda DNA templates up to 21 kbp
- Successfully amplifies GC-rich sequences
- Eliminates mispriming and primer-dimer formation
- Convenient room-temperature setup compatible with automation
- Optimal KOD Hot Start Buffer for PCR performance over wide range of targets
For Research Use Only