missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Novagen™ KOD Hot Start DNA Polymerase

For PCR amplification of long strand and GC-rich DNA templates, cloning and cDNA amplification applications

Brand:  EMD Millipore™ 71086-5

Code : Z3

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13148426

  • £68.67 / Each


Estimated Shipment
31-05-2018
Add to basket

Description

Description

  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA templates up to 21 kbp
  • Successfully amplifies GC-rich sequences
  • Eliminates mispriming and primer-dimer formation
  • Convenient room-temperature setup compatible with automation
  • Optimal KOD Hot Start Buffer for PCR performance over wide range of targets
Specifications

Specifications

1U/μL
Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli
KOD Hot Start DNA Polymerase
KOD Hot Start DNA Polymerase
For Accurate PCR Amplification
–20°C
20U
Videos

For Research Use Only