missing translation for 'onlineSavingsMsg'
Learn More

KLRG1 Mouse anti-Human, PE-eFluor™ 610, Clone: 13F12F2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Invitrogen 61-9488-41

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953324

  • £146.00 / Pack of 25
Estimated Shipment: 17-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This 13F12F2 monoclonal antibody reacts with human killer cell lectin-like receptor subfamily G, member 1 (KLRG1), a type II transmembrane inhibitor receptor of the C-type lectin superfamily that contains an ITIM domain. This inhibitory receptor is expressed on subsets of gamma-delta T cells, NK (CD56dim), CD8+ and CD4+ T Cells. KLRG1 is expressed primarily by cells with an effector/memory phenotype that are short-lived, but capable of immediate effector cell function. Cadherin-E, -N, and -R are ligands for KLRG1. Cadherin/KLRG1 interaction inhibits cytolytic activity and proliferation. The percentage of KLRG1 positive cells can vary considerably, depending on antigen experience. The clones 13F12F2 and 13A2 appear to recognize a similar epitope based on cross-blocking studies. Applications Reported: This 13F12F2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 13F12F2 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. KLRG1 is an inhibitory receptor that is expressed on natural killer (NK) cells and certain T cells. NK cells are involved in the lysis of tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.
Specifications

Specifications

KLRG1
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Q96E93
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, MAFA-like receptor, killer cell lectin-like receptor subfamily G member 1, mast cell function-associated antigen (ITIM-containing)
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
13F12F2
PE-eFluor 610
KLRG1
Liquid
KLRG1
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
10219
Videos
SDS
Documents
Certificates
Special Offers

Special Offers